Product Details

SNP ID
rs4847001
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:230648147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCCCAAAAATGTCACATAGTCC[C/T]TATGCTAAAGCACATGCATTTTATC
Phenotype
MIM: 606974
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COG2 PubMed Links

Gene Details

Gene
COG2
Gene Name
component of oligomeric golgi complex 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145036.1 Intron NP_001138508.1
NM_007357.2 Intron NP_031383.1

View Full Product Details