Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_175709.3 | 1552 | UTR 3 | NP_783640.1 | ||
XM_005261413.4 | 1552 | UTR 3 | XP_005261470.1 | ||
XM_006724174.3 | 1552 | UTR 3 | XP_006724237.1 | ||
XM_006724175.3 | 1552 | UTR 3 | XP_006724238.1 | ||
XM_006724176.3 | 1552 | UTR 3 | XP_006724239.1 | ||
XM_006724177.3 | 1552 | UTR 3 | XP_006724240.1 | ||
XM_006724178.3 | 1552 | UTR 3 | XP_006724241.1 | ||
XM_011530025.2 | 1552 | UTR 3 | XP_011528327.1 | ||
XM_017028679.1 | 1552 | UTR 3 | XP_016884168.1 |