Product Details

SNP ID
rs4300410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:76452107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAAGAGACTCCATCCCAGTTGTG[C/T]GCTCTGTTACAGAAAATTGTATAAT
Phenotype
MIM: 608574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMSY PubMed Links

Gene Details

Gene
EMSY
Gene Name
EMSY, BRCA2 interacting transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300942.1 Intron NP_001287871.1
NM_001300943.1 Intron NP_001287872.1
NM_001300944.1 Intron NP_001287873.1
NM_020193.4 Intron NP_064578.2
XM_005274112.4 Intron XP_005274169.1
XM_006718637.3 Intron XP_006718700.1
XM_006718638.3 Intron XP_006718701.1
XM_006718639.3 Intron XP_006718702.1
XM_011545173.2 Intron XP_011543475.1
XM_011545174.2 Intron XP_011543476.1
XM_011545175.2 Intron XP_011543477.1
XM_011545177.2 Intron XP_011543479.1
XM_011545178.2 Intron XP_011543480.1
XM_011545179.2 Intron XP_011543481.1
XM_017018021.1 Intron XP_016873510.1
XM_017018022.1 Intron XP_016873511.1

View Full Product Details