Product Details

SNP ID
rs4681969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57298932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAAGGTGCTTTGAATAAAATGA[G/T]GTCTCAATTCATATTTATGGAATGA
Phenotype
MIM: 603340
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASB14 PubMed Links

Gene Details

Gene
ASB14
Gene Name
ankyrin repeat and SOCS box containing 14
There are no transcripts associated with this gene.

Gene
DNAH12
Gene Name
dynein axonemal heavy chain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291661.1 Intron NP_001278590.1
NM_198564.3 Intron NP_940966.2
XM_011533466.1 Intron XP_011531768.1
XM_011533467.1 Intron XP_011531769.1
XM_011533468.1 Intron XP_011531770.1
XM_011533469.1 Intron XP_011531771.1
XM_011533471.2 Intron XP_011531773.1
XM_011533474.1 Intron XP_011531776.1
XM_017005860.1 Intron XP_016861349.1
XM_017005861.1 Intron XP_016861350.1
XM_017005862.1 Intron XP_016861351.1
XM_017005863.1 Intron XP_016861352.1

View Full Product Details