Product Details
- SNP ID
-
rs4912407
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.1:56647328 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTTTGACATTTGGACATGTACAC[A/G]CAAAGTTATATAACTTAACCCTATG
- Phenotype
-
MIM: 600497
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101929935
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75650857] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101929935
- Gene Name
- uncharacterized LOC101929935
There are no transcripts associated with this gene.
- Gene
- PRKAA2
- Gene Name
- protein kinase AMP-activated catalytic subunit alpha 2
View Full Product Details