Product Details

SNP ID
rs3918038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99482020 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGCCCAGACATCGTCTTTACGGA[C/G]CCGCTCCTGGTGGTTGATTTGGTTT
Phenotype
MIM: 603544 MIM: 615671
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCNK PubMed Links

Gene Details

Gene
CCNK
Gene Name
cyclin K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099402.1 Intron NP_001092872.1
XM_005268154.4 Intron XP_005268211.1
XM_011537275.2 Intron XP_011535577.1
Gene
SETD3
Gene Name
SET domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032233.2 Intron NP_115609.2
NM_199123.1 Intron NP_954574.1
XM_005268127.3 Intron XP_005268184.1
XM_011537231.2 Intron XP_011535533.2
XM_011537232.2 Intron XP_011535534.1
XM_011537233.2 Intron XP_011535535.1
XM_011537234.1 Intron XP_011535536.1
XM_011537235.1 Intron XP_011535537.1
XM_017021699.1 Intron XP_016877188.1
XM_017021700.1 Intron XP_016877189.1
XM_017021701.1 Intron XP_016877190.1
XM_017021702.1 Intron XP_016877191.1

View Full Product Details