Product Details

SNP ID
rs4602965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:75892338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCTTTCTGGTTCTCTTCCCCC[C/G]ACACCCCTGCGAGAAATGGGGGTAG
Phenotype
MIM: 600488
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PCSK5 PubMed Links

Gene Details

Gene
PCSK5
Gene Name
proprotein convertase subtilisin/kexin type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190482.1 Intron NP_001177411.1
NM_006200.5 Intron NP_006191.2
XM_005252039.3 Intron XP_005252096.1
XM_011518769.2 Intron XP_011517071.1
XM_011518770.2 Intron XP_011517072.1
XM_017014800.1 Intron XP_016870289.1

View Full Product Details