Product Details

SNP ID
rs6827221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109815870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGGAACCCAACTGCCGGAGGGAG[G/T]GAGGAAGGAAGGGGATGCGGTGCGA
Phenotype
MIM: 606468
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GAR1 PubMed Links

Gene Details

Gene
GAR1
Gene Name
GAR1 ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018983.3 Intron NP_061856.1
NM_032993.2 Intron NP_127460.1

View Full Product Details