Product Details
- SNP ID
-
rs4484654
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:39324669 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTCAGAAAATTTAGGGGAGAATAT[C/T]ATACAACCTGGATTTCTCAGTTGAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADAM5
PubMed Links
Gene Details
- Gene
- ADAM5
- Gene Name
- ADAM metallopeptidase domain 5 (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details