Product Details

SNP ID
rs6774551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194340701 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAGGCAGTGGCCTCCACTTTGT[A/C]TCACTGGGATGCAGTGAATAGACCT
Phenotype
MIM: 603104
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPN2 PubMed Links

Gene Details

Gene
CPN2
Gene Name
carboxypeptidase N subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080513.3 2092 UTR 3 NP_001073982.3
NM_001291988.1 2092 UTR 3 NP_001278917.1
XM_005269280.4 2092 UTR 3 XP_005269337.1

View Full Product Details