Product Details

SNP ID
rs7712080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:6717368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATGCAGTGTGTATTCCTCATG[A/G]GAGGATGAGCCTGCTGTGGAGCATT
Phenotype
MIM: 605198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100505625 PubMed Links

Gene Details

Gene
LOC100505625
Gene Name
uncharacterized LOC100505625
There are no transcripts associated with this gene.

Gene
PAPD7
Gene Name
PAP associated domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171805.1 Intron NP_001165276.1
NM_001171806.1 Intron NP_001165277.1
NM_006999.4 Intron NP_008930.1
XM_005248234.3 Intron XP_005248291.1
XM_011513945.1 Intron XP_011512247.1
XM_017008988.1 Intron XP_016864477.1
XM_017008989.1 Intron XP_016864478.1
XM_017008990.1 Intron XP_016864479.1

View Full Product Details