Product Details

SNP ID
rs7439465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:151673662 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATGACAACAACAAATCCCCAAA[C/T]TCATCAAACCTGAGAAAAAGGAAAC
Phenotype
MIM: 603645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM160A1 PubMed Links

Gene Details

Gene
FAM160A1
Gene Name
family with sequence similarity 160 member A1
There are no transcripts associated with this gene.

Gene
GATB
Gene Name
glutamyl-tRNA amidotransferase subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004564.2 Intron NP_004555.1
XM_005263063.3 Intron XP_005263120.1

View Full Product Details