Product Details
- SNP ID
-
rs52795588
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:26481832 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAAGCAGTAGATGATGGGGTTGA[C/T]GCAGGAGGAGGTGTAGGACAGGAGG
- Phenotype
-
MIM: 118444
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCKAR
PubMed Links
Gene Details
- Gene
- CCKAR
- Gene Name
- cholecystokinin A receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000730.2 |
1246 |
Missense Mutation |
ATC,GTC |
I365V |
NP_000721.1 |
View Full Product Details