Product Details
- SNP ID
-
rs7011501
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:74601353 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGTCTTTGTTCATGCTATTCTGT[A/G]TAAGAGAGTGTGCCCATAACATAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR2052HG
PubMed Links
Gene Details
- Gene
- MIR2052HG
- Gene Name
- MIR2052 host gene
There are no transcripts associated with this gene.
View Full Product Details