Product Details

SNP ID
rs7985521
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:73688917 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATAGGCGGTTTCACTTTCCTACT[C/T]TGCCGTTTACTATTTCTACATCCAT
Phenotype
MIM: 607531
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KLF12 PubMed Links

Gene Details

Gene
KLF12
Gene Name
Kruppel like factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007249.4 8113 UTR 3 NP_009180.3
XM_005266251.3 8113 UTR 3 XP_005266308.1
XM_011534907.2 8113 UTR 3 XP_011533209.1
XM_011534908.2 8113 Intron XP_011533210.1
XM_011534909.2 8113 UTR 3 XP_011533211.1
XM_011534910.2 8113 UTR 3 XP_011533212.1
XM_011534911.2 8113 UTR 3 XP_011533213.1
XM_011534912.2 8113 UTR 3 XP_011533214.1
XM_017020384.1 8113 UTR 3 XP_016875873.1
XM_017020385.1 8113 UTR 3 XP_016875874.1

View Full Product Details