Product Details

SNP ID
rs6735761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:149336511 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATGTGTCCACCAAGTTAAAGTCA[A/G]TGTTAGAAAAAGTGTAGGTTATAAA
Phenotype
MIM: 613359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYPD6 PubMed Links

Gene Details

Gene
LYPD6
Gene Name
LY6/PLAUR domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195685.1 Intron NP_001182614.1
NM_194317.3 Intron NP_919298.1

View Full Product Details