Product Details
- SNP ID
-
rs6792143
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51818569 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTGTGCACATGAGTCCACTTATGT[A/G]TAGGTAAGCTGAAGTTAAAAGAAGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IQCF3
PubMed Links
Gene Details
- Gene
- IQCF3
- Gene Name
- IQ motif containing F3
There are no transcripts associated with this gene.
- Gene
- IQCF4
- Gene Name
- IQ motif containing F5 pseudogene
There are no transcripts associated with this gene.
View Full Product Details