Product Details
- SNP ID
-
rs7582672
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:207717492 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTTACATTCTGGAGGGAGCTAGG[A/G]ATAGATAATACATGAATAATCAAGA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCNYL1
PubMed Links
Gene Details
- Gene
- CCNYL1
- Gene Name
- cyclin Y like 1
View Full Product Details