Product Details

SNP ID
rs7008370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33593567 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTCAGCGCCCGGTGGATGAAGTC[A/G]GCAGCCGTCTGGAAGTGGATGCTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP26 PubMed Links

Gene Details

Gene
DUSP26
Gene Name
dual specificity phosphatase 26 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305115.1 684 Silent Mutation GCC,GCT A134A NP_001292044.1
NM_001305116.1 684 Silent Mutation GCC,GCT A134A NP_001292045.1
NM_024025.2 684 Silent Mutation GCC,GCT A134A NP_076930.1

View Full Product Details