Product Details
- SNP ID
-
rs7982594
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:51338097 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTAAAAGCAAGTTAGAATGGCATA[G/T]TCATAGAGTTAGAAAGAAATGTTTG
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SERPINE3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs137951517,rs79458946] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SERPINE3
- Gene Name
- serpin family E member 3
View Full Product Details