Product Details
- SNP ID
-
rs6744089
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218711880 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTTCTCAGATACTACAAATCACCT[A/G]TTCTTTAACTGCAGCACCTGTTGAT
- Phenotype
-
MIM: 607652
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
STK36
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80102405] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- STK36
- Gene Name
- serine/threonine kinase 36
There are no transcripts associated with this gene.
- Gene
- TTLL4
- Gene Name
- tubulin tyrosine ligase like 4
View Full Product Details