Product Details
- SNP ID
-
hCV29018888
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218615745 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGGTGGAACAGTGGGTGATGGC[A/G]TCCCTGCTGCAAGACCGTGAGTGCC
- Phenotype
-
MIM: 605939
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLCD4
PubMed Links
Gene Details
- Gene
- PLCD4
- Gene Name
- phospholipase C delta 4
There are no transcripts associated with this gene.
View Full Product Details