Product Details

SNP ID
hCV29023014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47249160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAGCGTCCCCTCCAGGCTTGAC[C/T]CCATAGCTAGATCGGGTGCAGAATT
Phenotype
MIM: 171650 MIM: 600811 MIM: 602423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACP2 PubMed Links

Gene Details

Gene
ACP2
Gene Name
acid phosphatase 2, lysosomal
There are no transcripts associated with this gene.

Gene
DDB2
Gene Name
damage specific DNA binding protein 2
There are no transcripts associated with this gene.

Gene
NR1H3
Gene Name
nuclear receptor subfamily 1 group H member 3
There are no transcripts associated with this gene.

View Full Product Details