Product Details
- SNP ID
-
rs7598273
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:74138358 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTAGCATTAAGTCCAACACAAATG[A/G]GCTGAATGTGGGCAGCAGAGGACTG
- Phenotype
-
MIM: 613183
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BOLA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113284534] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BOLA3
- Gene Name
- bolA family member 3
- Gene
- BOLA3-AS1
- Gene Name
- BOLA3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details