Product Details

SNP ID
rs7696518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5543521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAGTATACAGAGTATACCACAAA[C/T]AACAAAATTGGCAAAAATACATTAC
Phenotype
MIM: 607261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EVC2 PubMed Links

Gene Details

Gene
EVC2
Gene Name
EvC ciliary complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166136.1 Intron NP_001159608.1
NM_147127.4 Intron NP_667338.3
XM_017007735.1 Intron XP_016863224.1
XM_017007736.1 Intron XP_016863225.1
XM_017007737.1 Intron XP_016863226.1
XM_017007738.1 Intron XP_016863227.1
XM_017007739.1 Intron XP_016863228.1

View Full Product Details