Product Details
- SNP ID
-
rs7185401
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:69201014 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCGTTTATATGTCACTGAAAACTC[A/G]GACAAATCGAGTCATTTCTCTGCTT
- Phenotype
-
MIM: 600027
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNTB2
PubMed Links
Gene Details
- Gene
- SNTB2
- Gene Name
- syntrophin beta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006750.3 |
|
Intron |
|
|
NP_006741.1 |
View Full Product Details