Product Details

SNP ID
rs6863749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:61161800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCCACGATTTTCAACTGAACA[C/T]ACATGAAAGTGCCTAATACGTTAAA
Phenotype
MIM: 609653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTC-436P18.1 PubMed Links

Gene Details

Gene
CTC-436P18.1
Gene Name
uncharacterized LOC101928630
There are no transcripts associated with this gene.

Gene
NDUFAF2
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 2
There are no transcripts associated with this gene.

Gene
SMIM15
Gene Name
small integral membrane protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048249.3 Intron NP_001041714.1

View Full Product Details