Product Details

SNP ID
rs8044820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87402776 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCATGTCAATATAATCAAAGGAA[A/G]TGTCCTGTGCTTTATGAGAAGTATT
Phenotype
MIM: 609604
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP1LC3B PubMed Links

Gene Details

Gene
MAP1LC3B
Gene Name
microtubule associated protein 1 light chain 3 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022818.4 Intron NP_073729.1
Gene
ZCCHC14
Gene Name
zinc finger CCHC-type containing 14
There are no transcripts associated with this gene.

View Full Product Details