Product Details

SNP ID
rs6674171
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:154519207 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCGTTCTCACGAATTGCTTGTTC[A/G]GCATCTTTTAGATTCATGTGTAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDRD10 PubMed Links

Gene Details

Gene
TDRD10
Gene Name
tudor domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098475.1 Intron NP_001091945.1
NM_182499.3 Intron NP_872305.3
XM_006711156.2 Intron XP_006711219.1
XM_011509152.1 Intron XP_011507454.1
XM_011509153.1 Intron XP_011507455.1
XM_011509154.1 Intron XP_011507456.1
XM_011509155.1 Intron XP_011507457.1
XM_011509156.1 Intron XP_011507458.1
XM_011509157.1 Intron XP_011507459.1
XM_011509158.2 Intron XP_011507460.1
XM_011509159.2 Intron XP_011507461.1
XM_011509160.2 Intron XP_011507462.1
XM_011509161.1 Intron XP_011507463.1
XM_011509162.2 Intron XP_011507464.1
XM_017000255.1 Intron XP_016855744.1

View Full Product Details