Product Details

SNP ID
rs7505388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:56651431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCACTGGGGAGACGGCGGCTGTA[C/T]AGCGCTTGCCGCCCCACGGATTATC
Phenotype
MIM: 603049 MIM: 613473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNL1 PubMed Links

Gene Details

Gene
TXNL1
Gene Name
thioredoxin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004786.2 47 Intron NP_004777.1
XM_006722580.3 47 Intron XP_006722643.1
XM_011526263.2 47 Intron XP_011524565.1
XM_011526264.2 47 Intron XP_011524566.1
XM_017026093.1 47 Intron XP_016881582.1
XM_017026094.1 47 Intron XP_016881583.1
Gene
WDR7
Gene Name
WD repeat domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015285.2 47 UTR 5 NP_056100.2
NM_052834.2 47 UTR 5 NP_443066.2
XM_006722431.2 47 Intron XP_006722494.1
XM_011525888.1 47 UTR 5 XP_011524190.1
XM_017025682.1 47 Intron XP_016881171.1
XM_017025683.1 47 Intron XP_016881172.1

View Full Product Details