Product Details

SNP ID
rs6987438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80634836 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATCTTTTGGAAATGAGACTCATT[G/T]GGTTCAAAACCGTACAGCAATAAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF704 PubMed Links

Gene Details

Gene
ZNF704
Gene Name
zinc finger protein 704
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033723.2 8632 UTR 3 NP_001028895.1
XM_005251279.4 8632 UTR 3 XP_005251336.2
XM_017013725.1 8632 UTR 3 XP_016869214.1

View Full Product Details