Product Details

SNP ID
rs7776527
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2687703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGGGCCTGGTTCACCCCTCTCAC[C/T]TGGGGCTTCGCTGATAAGCCTGGTT
Phenotype
MIM: 615168
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AMZ1 PubMed Links

Gene Details

Gene
AMZ1
Gene Name
archaelysin family metallopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284355.2 Intron NP_001271284.1
NM_001321766.1 Intron NP_001308695.1
NM_133463.3 Intron NP_597720.1
XM_011515150.2 Intron XP_011513452.1
XM_011515151.2 Intron XP_011513453.1
XM_017011774.1 Intron XP_016867263.1
XM_017011775.1 Intron XP_016867264.1
XM_017011776.1 Intron XP_016867265.1

View Full Product Details