Product Details

SNP ID
rs8076826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:65011804 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGCATAGTGAAATTATGAGCAC[C/T]TTTTCAATTCTGTTCACTAAATTTC
Phenotype
MIM: 604406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNA13 PubMed Links

Gene Details

Gene
GNA13
Gene Name
G protein subunit alpha 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282425.1 3486 UTR 3 NP_001269354.1
NM_006572.5 3486 UTR 3 NP_006563.2
XM_011524202.1 3486 Intron XP_011522504.1

View Full Product Details