Product Details

SNP ID
rs7839394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109541365 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAATTAAAAGGTTTCTTAAGTA[A/G]GATATGATGTAGGGAGAGCCTTGGG
Phenotype
MIM: 605772 MIM: 607843
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EBAG9 PubMed Links

Gene Details

Gene
EBAG9
Gene Name
estrogen receptor binding site associated, antigen, 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278938.1 Intron NP_001265867.1
NM_004215.4 Intron NP_004206.1
NM_198120.2 Intron NP_936056.1
XM_017013960.1 Intron XP_016869449.1
Gene
PKHD1L1
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1
There are no transcripts associated with this gene.

View Full Product Details