Product Details

SNP ID
rs7718623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:82290644 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTGCTCCTTGGGGAATGGAGG[A/G]TGCTATGAACTGATGTAACCTGGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6AP1L PubMed Links

Gene Details

Gene
ATP6AP1L
Gene Name
ATPase H+ transporting accessory protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017971.2 Intron NP_001017971.1
XM_011543719.2 Intron XP_011542021.1
XM_011543720.2 Intron XP_011542022.1
XM_011543722.2 Intron XP_011542024.1
XM_011543723.2 Intron XP_011542025.1
XM_011543724.2 Intron XP_011542026.1
XM_017010025.1 Intron XP_016865514.1
XM_017010026.1 Intron XP_016865515.1
XM_017010027.1 Intron XP_016865516.1
XM_017010028.1 Intron XP_016865517.1
XM_017010029.1 Intron XP_016865518.1
XM_017010030.1 Intron XP_016865519.1
XM_017010031.1 Intron XP_016865520.1
XM_017010032.1 Intron XP_016865521.1
XM_017010033.1 Intron XP_016865522.1
XM_017010034.1 Intron XP_016865523.1
XM_017010035.1 Intron XP_016865524.1
XM_017010036.1 Intron XP_016865525.1
XM_017010037.1 Intron XP_016865526.1
XM_017010038.1 Intron XP_016865527.1
XM_017010039.1 Intron XP_016865528.1
XM_017010040.1 Intron XP_016865529.1
XM_017010041.1 Intron XP_016865530.1
XM_017010042.1 Intron XP_016865531.1
XM_017010043.1 Intron XP_016865532.1
XM_017010044.1 Intron XP_016865533.1
XM_017010045.1 Intron XP_016865534.1
XM_017010046.1 Intron XP_016865535.1
XM_017010047.1 Intron XP_016865536.1

View Full Product Details