Product Details

SNP ID
rs4831971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:84429329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCCAAATACATTTTAAATTCAA[C/T]TTAACACAGCACAATGCTGTATTAA
Phenotype
MIM: 611224
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUCLG1 PubMed Links

Gene Details

Gene
SUCLG1
Gene Name
succinate-CoA ligase alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003849.3 Intron NP_003840.2

View Full Product Details