Product Details
- SNP ID
-
rs4402279
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:1476554 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGGCTTACCAATTTATTACCTTTC[A/C]AAGTCATCACATTTTGATTTCACTC
- Phenotype
-
MIM: 611969
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MOB2
PubMed Links
Gene Details
- Gene
- MOB2
- Gene Name
- MOB kinase activator 2
View Full Product Details