Product Details

SNP ID
rs7619423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126438587 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTGACTTCTCTTACTCAAAACTA[C/T]AGGCTCCCCTTATTCTAAACACATG
Phenotype
MIM: 615746
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP100 PubMed Links

Gene Details

Gene
CFAP100
Gene Name
cilia and flagella associated protein 100
There are no transcripts associated with this gene.

Gene
LOC105374089
Gene Name
uncharacterized LOC105374089
There are no transcripts associated with this gene.

Gene
ZXDC
Gene Name
ZXD family zinc finger C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040653.3 Intron NP_001035743.1
NM_025112.4 Intron NP_079388.3
XM_005247757.3 Intron XP_005247814.1
XM_006713741.2 Intron XP_006713804.1
XM_011513119.2 Intron XP_011511421.1

View Full Product Details