Product Details

SNP ID
rs3187463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:71187515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGTGCATAAACTTGTCTGAGGG[A/G]ATTAAGACACCAGGAAGACCAAGTG
Phenotype
MIM: 611018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAIP2B PubMed Links

Gene Details

Gene
PAIP2B
Gene Name
poly(A) binding protein interacting protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020459.1 1493 UTR 3 NP_065192.1
XM_005264310.3 1493 UTR 3 XP_005264367.1
XM_005264311.4 1493 UTR 3 XP_005264368.1
XM_011532842.2 1493 UTR 3 XP_011531144.1
XM_011532843.2 1493 UTR 3 XP_011531145.1

View Full Product Details