Product Details

SNP ID
rs7690706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102875592 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCCATGAACAAAGACAAGTAAGT[A/C]CAGACCTTGTAGGAATTATCTCAAA
Phenotype
MIM: 611507 MIM: 611527 MIM: 602963
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CISD2 PubMed Links

Gene Details

Gene
CISD2
Gene Name
CDGSH iron sulfur domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008388.4 Intron NP_001008389.1
Gene
SLC9B1
Gene Name
solute carrier family 9 member B1
There are no transcripts associated with this gene.

Gene
UBE2D3
Gene Name
ubiquitin conjugating enzyme E2 D3
There are no transcripts associated with this gene.

View Full Product Details