Product Details

SNP ID
rs6533206
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:105552939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAGGAGCCAATCTGACAGGAC[C/T]GAATACAACCAGAAATTACAAGGTA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ARHGEF38 PubMed Links

Gene Details

Gene
ARHGEF38
Gene Name
Rho guanine nucleotide exchange factor 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242729.1 57 Silent Mutation ACC,ACT T58T NP_001229658.1
NM_017700.1 57 Silent Mutation ACC,ACT T58T NP_060170.1
XM_006714244.3 57 Intron XP_006714307.1
XM_011532050.2 57 UTR 5 XP_011530352.1
XM_011532051.2 57 Silent Mutation ACC,ACT T58T XP_011530353.1
Gene
ARHGEF38-IT1
Gene Name
ARHGEF38 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details