Product Details
- SNP ID
-
rs7921703
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:109999361 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCTCAGAAAGCTCACATGCATAC[A/G]CTTCCTCCTGCCACCGCCATCACCT
- Phenotype
-
MIM: 601568
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADD3
PubMed Links
Gene Details
- Gene
- ADD3
- Gene Name
- adducin 3
- Gene
- ADD3-AS1
- Gene Name
- ADD3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details