Product Details
- SNP ID
-
rs7780789
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:121328474 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATAAATGCATACATAAATGACAG[C/T]GATTGTAAGCCTGACAGCACTTCAC
- Phenotype
-
MIM: 606267
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WNT16
PubMed Links
Gene Details
- Gene
- WNT16
- Gene Name
- Wnt family member 16
View Full Product Details