Product Details

SNP ID
rs6912288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39190005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCGGGCAGGGTGTGCAGTTCTC[A/C]GGCGGGCAGGAGGGGTAAGGGCAGA
Phenotype
MIM: 603493
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNK5 PubMed Links

Gene Details

Gene
KCNK5
Gene Name
potassium two pore domain channel subfamily K member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003740.3 2749 UTR 3 NP_003731.1
XM_005249456.1 2749 UTR 3 XP_005249513.1
XM_006715235.2 2749 UTR 3 XP_006715298.1

View Full Product Details