Product Details

SNP ID
rs7069846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71219869 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAGGCATGGGGCTTCCTGGGGC[A/G]GGTGATGAGTGCCAGACCTTGACTG
Phenotype
MIM: 607870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UNC5B PubMed Links

Gene Details

Gene
UNC5B
Gene Name
unc-5 netrin receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244889.1 Intron NP_001231818.1
NM_170744.4 Intron NP_734465.2
XM_011539453.1 Intron XP_011537755.1
XM_017015834.1 Intron XP_016871323.1
XM_017015835.1 Intron XP_016871324.1
Gene
UNC5B-AS1
Gene Name
UNC5B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details