Product Details

SNP ID
rs7251136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12752150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGGGGAAGTGGAGAAGGGGGA[C/G]CCAGAAATAGGGGCTGAGGGAGAGA
Phenotype
MIM: 601913 MIM: 607335
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASNA1 PubMed Links

Gene Details

Gene
ASNA1
Gene Name
arsA arsenite transporter, ATP-binding, homolog 1 (bacterial)
There are no transcripts associated with this gene.

Gene
BEST2
Gene Name
bestrophin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017682.2 Intron NP_060152.2
XM_005259963.3 Intron XP_005260020.1

View Full Product Details