Product Details

SNP ID
rs7503669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7560142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATATGGTGTCCGAATCCAGGATG[C/T]TGGAGTTTATCTGCTGTATAGCCAG
Phenotype
MIM: 612844 MIM: 602695 MIM: 604472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SENP3 PubMed Links

Gene Details

Gene
SENP3
Gene Name
SUMO1/sentrin/SMT3 specific peptidase 3
There are no transcripts associated with this gene.

Gene
SENP3-EIF4A1
Gene Name
SENP3-EIF4A1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TNFSF12
Gene Name
tumor necrosis factor superfamily member 12
There are no transcripts associated with this gene.

Gene
TNFSF12-TNFSF13
Gene Name
TNFSF12-TNFSF13 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172089.3 1227 Missense Mutation GCT,GTT A240V NP_742086.1
Gene
TNFSF13
Gene Name
tumor necrosis factor superfamily member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198622.1 1227 Missense Mutation GCT,GTT A133V NP_001185551.1
NM_001198623.1 1227 Missense Mutation GCT,GTT A132V NP_001185552.1
NM_001198624.1 1227 Missense Mutation GCT,GTT A115V NP_001185553.1
NM_003808.3 1227 Missense Mutation GCT,GTT A160V NP_003799.1
NM_172087.2 1227 Missense Mutation GCT,GTT A144V NP_742084.1
NM_172088.2 1227 Missense Mutation GCT,GTT A160V NP_742085.1

View Full Product Details