Product Details

SNP ID
rs8065733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41167233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAAGAGATGAGCATTTTTGGTG[C/T]TTTTGAACACTATGAAGTTTTATTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP4-3 PubMed Links

Gene Details

Gene
KRTAP4-3
Gene Name
keratin associated protein 4-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033187.1 940 UTR 3 NP_149443.1
Gene
KRTAP4-4
Gene Name
keratin associated protein 4-4
There are no transcripts associated with this gene.

View Full Product Details