Product Details

SNP ID
rs6521125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46582440 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACATAGATTTGATTTCATTTTTTC[A/G]TCTTGGCCAGTTTGATGAATGGAAA
Phenotype
MIM: 300375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHST7 PubMed Links

Gene Details

Gene
CHST7
Gene Name
carbohydrate sulfotransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019886.3 Intron NP_063939.2

View Full Product Details