Product Details

SNP ID
rs8100539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33631036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGCCCAGTGTGCAATGCAGTCTG[C/T]CTACACTAGTTGGAATCAGTATGTG
Phenotype
MIM: 610190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST8 PubMed Links

Gene Details

Gene
CHST8
Gene Name
carbohydrate sulfotransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127895.1 Intron NP_001121367.1
NM_001127896.1 Intron NP_001121368.1
NM_022467.3 Intron NP_071912.2
XM_011527222.1 Intron XP_011525524.1
XM_011527224.1 Intron XP_011525526.1
XM_017027143.1 Intron XP_016882632.1

View Full Product Details